Order Entry
Norway
Orders LinkContactUsLinkComponent
Boc-Pro-OH

Boc-Pro-OH

Irritant
Supplier:   Apollo Scientific
Boc-Pro-OH
Supplier:  Apollo Scientific
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid
  • CAS Number:15761-39-4
  • Formula:C₁₀H₁₇NO₄
  • MDL Number:MFCD00037324
  • Molecular Weight:215.25 g/mol
  • Melting Pt:133…136 °C
  • Storage Temperature:Refrigerator
  • EINECS:239-848-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - kr 270,00
    kr 270,00
    1 * 5 g
       kr 270,00

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg - kr 31 400,00
    kr 31 400,00
    1 * 25 kg
       kr 31 400,00

    Specifications:
    New Product
    New Product