Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2-methylbenzaldehyde
4-Bromo-2-methylbenzaldehyde
4-Bromo-2-methylbenzaldehyde
4-Bromo-2-methylbenzaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
4-Bromo-2-methylbenzaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
24078-12-4
:
Formula:
C₈H₇BrO
MDL Number:
MFCD07787171
:
199.05
:
GHS07
Storage Temperature:
Ambient