Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-cyanobenzoate
Methyl-4-cyanobenzoate 98%
Methyl-4-cyanobenzoate 98%
Methyl-4-cyanobenzoate 98%
APOSOR5721-100G
:
Apollo Scientific
:
Methyl-4-cyanobenzoate 98%
APOSOR5721-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
161.16 g/mol
:
142…144 °C/12mmHg
:
65…69 °C
:
214-443-4
:
GHS07
:
Ambient
:
1129-35-7
:
C₉H₇NO₂
:
MFCD00001823