
3,5-Dichlorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:705-21-5
:
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00051698
:245.51 g/mol
Boiling Pt:85…86 °C/0.5mmHg
Melting Pt:30…32 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05