Order Entry
Norway
ContactUsLinkComponent
Sulpholane 99%
Irritant
Sulpholane 99%
Catalog # APOSOR51759-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Sulpholane 99%
Catalog # APOSOR51759-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :120.17 g/mol
EINECS:204-783-1
 :GHS07
Storage Temperature:Ambient
CAS Number:126-33-0
Formula:C₄H₈O₂S
MDL Number:MFCD00005484