

3-Phenylpropylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2038-57-5
:
Formula:C₉H₁₃N
MDL Number:MFCD00008224
:135.21 g/mol
Boiling Pt:220…221 °C
Density:0.949 g/cm³ (25 °C)
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:218-012-1
UN:2735
ADR:8,III
:GHS05
:GHS07