4-(Bromomethyl)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:66176-39-4
:
Formula:C₇H₆BrClO₂S
MDL Number:MFCD00156129
:269.55
Melting Pt:73…76 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05