kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
4-Formylbenzoic acid
4-Formylbenzoic acid
4-Formylbenzoic acid
Irritant
4-Formylbenzoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Formylbenzoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
619-66-9
:
Formula:
C₈H₆O₃
MDL Number:
MFCD00006951
:
150.13 g/mol
Melting Pt:
245…247 °C
EINECS:
210-607-4
:
GHS07
Storage Temperature:
Ambient