kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
(3-Nitrophenyl)acetic acid
(3-Nitrophenyl)acetic acid
(3-Nitrophenyl)acetic acid
Irriterende
(3-Nitrophenyl)acetic acid
Leverandør:
Apollo Scientific
Sertifikat
Utvid bildet
Irriterende
(3-Nitrophenyl)acetic acid
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
1877-73-2
:
Formel:
(C₆H₄(NO₂))CH₂COOH
MDL-nummer:
MFCD00007278
:
181.15 g/mol
Molten Pt:
117…120 °C
:
GHS07
Lagringstemperatur:
Ambient