Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Isoquinolinol
4-Isoquinolinol
4-Isoquinolinol
Irritant
4-Isoquinolinol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Isoquinolinol
Supplier:
Apollo Scientific
About this item
CAS Number:
3336-49-0
:
Formula:
C₉H₇NO
MDL Number:
MFCD00234514
:
145.1580 g/mol
:
GHS07
Storage Temperature:
Refrigerator