Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Dimethyldihydro-2H-pyran-4(3H)-one
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
Irritant
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,2-Dimethyldihydro-2H-pyran-4(3H)-one 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1194-16-7
:
Formula:
C₇H₁₂O₂
MDL Number:
MFCD01549337
:
128.17 g/mol
Boiling Pt:
60…64 °C/8mmHg
:
GHS07
Storage Temperature:
Refrigerator