Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methoxy-2-methylbenzothiazole
6-Methoxy-2-methylbenzothiazole 97%
6-Methoxy-2-methylbenzothiazole 97%
6-Methoxy-2-methylbenzothiazole 97%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Methoxy-2-methylbenzothiazole 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2941-72-2
:
Formula:
C₉H₉NOS
MDL Number:
MFCD00005795
:
179.24 g/mol
Storage Temperature:
Ambient