tert-Butyl-5-oxo-D-prolinate
Catalog # APOSOR46095-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:185.22 g/mol
Storage Temperature:Ambient
CAS Number:205524-46-5
Formula:C₉H₁₅NO₃
MDL Number:MFCD09261417