Order Entry
Norway
ContactUsLinkComponent
3-Bromo-p-anisaldehyde 95%
3-Bromo-p-anisaldehyde 95%
Katalog # APOSOR4599-100G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-p-anisaldehyde 95%
Katalog # APOSOR4599-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :215.05 g/mol
Melting Pt:51…53 °C
Storage Temperature:Ambient
CAS Number:34841-06-0
Formula:C₈H₇BrO₂
MDL Number:MFCD00016599