Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cinnamamide
Cinnamamide 97%
Cinnamamide 97%
Irritant
Cinnamamide 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Cinnamamide 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
621-79-4
:
Formula:
C₉H₉NO
MDL Number:
MFCD00008033
:
147.177 g/mol
Melting Pt:
148…150 °C
:
GHS07
Storage Temperature:
Ambient