Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
p-Tolylacetic acid
p-Tolylacetic acid 98%
p-Tolylacetic acid 98%
:
Apollo Scientific
p-Tolylacetic acid 98%
:
Apollo Scientific
:
622-47-9
:
:
(C₆H₄(CH₃))CH₂COOH
:
MFCD00004353
:
150.18 g/mol
:
265…267 °C
:
88…92 °C
:
Ambient
:
210-738-7
:
GHS07