

1-Boc-2-piperidone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85908-96-9
:
Formula:C₁₀H₁₇NO₃
MDL Number:MFCD02179046
:199.25 g/mol
Melting Pt:34…37 °C
Flash Pt:110 °C
Storage Temperature:Romtemperatur
UN:3082
ADR:9+EHS,III
:GHS07
:GHS09