8-Bromoquinoline-3-carboxylic acid
Catalog # APOSOR42249-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.0641 g/mol
Storage Temperature:Ambient
CAS Number:347146-16-1
Formula:C₁₀H₆BrNO₂
MDL Number:MFCD18375331