Order Entry
Norway
ContactUsLinkComponent
2-Amino-5-chloro-4-methylpyridine 95+%
2-Amino-5-chloro-4-methylpyridine 95+%
Catalog # APOSOR40192-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-5-chloro-4-methylpyridine 95+%
Catalog # APOSOR40192-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :142.5900 g/mol
Storage Temperature:Ambient
CAS Number:36936-27-3
Formula:C₆H₇ClN₂
MDL Number:MFCD06410759