
2,6-Dimethyl-4-pyridineboronic acid
Catalog # APOSOR361167-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:150.9708 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:846548-44-5
Formula:C₇H₁₀BNO₂
MDL Number:MFCD11042425