


3-Chloro-o-toluidine
Catalog # APOSOR350077-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:141.6 g/mol
Boiling Pt:115…117 °C
Density:1.185 g/cm³ (25 °C)
Flash Pt:113 °C
Storage Temperature:Ambient
UN:3429
ADR:6.1,III
:GHS05
:GHS07
:GHS08
CAS Number:87-60-5
Formula:C₇H₈ClN
MDL Number:MFCD00007766