Order Entry
Norway
ContactUsLinkComponent
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide 95%
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide 95%
Catalog # APOSOR33558-1G
Supplier:  Apollo Scientific
CAS Number:  
N,4,5-Trimethyl-1,3-thiazol-2-amine hydrobromide 95%
Catalog # APOSOR33558-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.14 g/mol
Storage Temperature:Ambient
CAS Number:849004-10-0
Formula:C₆H₁₁BrN₂S
MDL Number:MFCD18651784