Order Entry
Norway
ContactUsLinkComponent
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol 95%
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol 95%
Catalog # APOSOR32618-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-Methyl-3-phenyltetrahydro-5-isoxazolyl)methanol 95%
Catalog # APOSOR32618-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.25 g/mol
Storage Temperature:Ambient
CAS Number:19345-02-9
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD00171848