Margaric acid
Catalog # APOSOR322165-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:270.46 g/mol
Boiling Pt:227 °C/100mmHg
Melting Pt:59…61 °C
Density:0.853 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:208-027-1
:GHS07
CAS Number:506-12-7
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00002751