Order Entry
Norway
ContactUsLinkComponent
Ethyl-2-bromoisovalerate 98%
Ethyl-2-bromoisovalerate 98%
Catalog # APOSOR322033-25G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-bromoisovalerate 98%
Catalog # APOSOR322033-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :209.08 g/mol
Storage Temperature:Ambient
CAS Number:609-12-1
Formula:C₇H₁₃BrO₂
MDL Number:MFCD00026855