Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromothieno[3,2-b]thiophene
3-Bromothieno[3,2-b]thiophene
3-Bromothieno[3,2-b]thiophene
APOSOR310409-1G
:
Apollo Scientific
:
3-Bromothieno[3,2-b]thiophene
APOSOR310409-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
219.11 g/mol
:
1.78 g/cm³ (25 °C)
:
GHS07
:
Refrigerator
:
25121-83-9
:
C₆H₃BrS₂
:
MFCD10000952