Order Entry
Norway
ContactUsLinkComponent
5-Methylisatin 98%
Irritant
5-Methylisatin 98%
Catalog # APOSOR30934-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methylisatin 98%
Catalog # APOSOR30934-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :161.16 g/mol
Melting Pt:180 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:608-05-9
Formula:C₉H₇NO₂
MDL Number:MFCD00005721