Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-bromophenol
2-Amino-5-bromophenol 98%
2-Amino-5-bromophenol 98%
Irritant
2-Amino-5-bromophenol 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-5-bromophenol 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
38191-34-3
:
Formula:
C₆H₆BrNO
MDL Number:
MFCD03095028
:
188.0200 g/mol
Melting Pt:
135…140 °C (dec.)
:
GHS07
Storage Temperature:
Ambient