3-(4-Bromophenyl)propionic acid
Catalog # APOSOR3085-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:229.07 g/mol
Storage Temperature:Ambient
CAS Number:1643-30-7
Formula:C₉H₉BrO₂
MDL Number:MFCD01310793