Order Entry
Norway
ContactUsLinkComponent
2,3-Dimethylbenzoic acid 97%
Irritant
2,3-Dimethylbenzoic acid 97%
Catalog # APOSOR30294-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Dimethylbenzoic acid 97%
Catalog # APOSOR30294-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :150.18 g/mol
Melting Pt:144…146 °C
EINECS:210-058-0
 :GHS07
Storage Temperature:Refrigerator
CAS Number:603-79-2
Formula:C₉H₁₀O₂
MDL Number:MFCD00002479