Order Entry
Norway
ContactUsLinkComponent
Methyl-5-methoxynicotinate
Methyl-5-methoxynicotinate
Catalog # APOSOR302228-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-5-methoxynicotinate
Catalog # APOSOR302228-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.16 g/mol
Storage Temperature:Ambient
CAS Number:29681-46-7
Formula:C₈H₉NO₃
MDL Number:MFCD09265109