Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-6-methylpyrimidine
4-Hydroxy-6-methylpyrimidine 97+%
4-Hydroxy-6-methylpyrimidine 97+%
Corrosive
Irritant
4-Hydroxy-6-methylpyrimidine 97+%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
4-Hydroxy-6-methylpyrimidine 97+%
Supplier:
Apollo Scientific
About this item
CAS Number:
3524-87-6
:
Formula:
C₅H₆N₂O
MDL Number:
MFCD00053575
:
110.116 g/mol
Melting Pt:
145…150 °C
:
GHS05
:
GHS07
Storage Temperature:
Ambient