kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
2-Bromo-4-methylbenzaldehyde
2-Bromo-4-methylbenzaldehyde 98%
2-Bromo-4-methylbenzaldehyde 98%
APOSOR301009-25G
:
Apollo Scientific
:
2-Bromo-4-methylbenzaldehyde 98%
APOSOR301009-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
199.05
:
30…35 °C
:
>110 °C
:
GHS07
:
Refrigerator
:
824-54-4
:
C₈H₇BrO
:
MFCD04039890