5-(Chloromethyl)quinolin-8-ol hydrochloride
Catalog # APOSOR2892-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.1 g/mol
Storage Temperature:Ambient
CAS Number:4053-45-6
Formula:C₁₀H₉Cl₂NO
MDL Number:MFCD01790514