Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nitrocinnamaldehyde
2-Nitrocinnamaldehyde
2-Nitrocinnamaldehyde
APOSOR28735-25G
:
Apollo Scientific
:
2-Nitrocinnamaldehyde
APOSOR28735-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
127…129 °C
:
177.16 g/mol
:
215-988-0
:
GHS07
:
Refrigerator
:
1466-88-2
:
C₉H₇NO₃
:
MFCD00007188