Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloroisonicotinic acid
2,3-Dichloroisonicotinic acid
2,3-Dichloroisonicotinic acid
Irritant
2,3-Dichloroisonicotinic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Dichloroisonicotinic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
184416-84-0
:
Formula:
C₆H₃Cl₂NO₂
MDL Number:
MFCD01861977
:
192 g/mol
Melting Pt:
187 °C
:
GHS07
Storage Temperature:
Ambient