Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
2,5-Dichloro-4-pyridineboronic acid
2,5-Dichloro-4-pyridineboronic acid
2,5-Dichloro-4-pyridineboronic acid
Irritant
2,5-Dichloro-4-pyridineboronic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,5-Dichloro-4-pyridineboronic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
847664-64-6
:
Formula:
C₅H₄BCl₂NO₂
MDL Number:
MFCD06798254
:
191.81 g/mol
Melting Pt:
161…163 °C
:
GHS07
Storage Temperature:
Romtemperatur