Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Iodo-3-pyridinol
2-Iodo-3-pyridinol
2-Iodo-3-pyridinol
2-Iodo-3-pyridinol
Supplier:
Apollo Scientific
Certificates
Expand image
2-Iodo-3-pyridinol
Supplier:
Apollo Scientific
About this item
CAS Number:
40263-57-8
:
Formula:
C₅H₄INO
MDL Number:
MFCD00023421
:
221.00 g/mol
Melting Pt:
196…200 °C
EINECS:
254-864-0
:
GHS07
Storage Temperature:
Ambient