ω-Bromo-4-chloroacetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:536-38-9
:
Formula:C₈H₆BrClO
MDL Number:MFCD00000625
:233.49 g/mol
Melting Pt:93…98 °C
Storage Temperature:Ambient
EINECS:208-631-5
UN:3261
ADR:8,II
:GHS05