Order Entry
Norway
ContactUsLinkComponent
2-Iodomesitylene 98%
2-Iodomesitylene 98%
Catalog # APOSOR23747-100G
Supplier:  Apollo Scientific
CAS Number:  
2-Iodomesitylene 98%
Catalog # APOSOR23747-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :246.09 g/mol
Melting Pt:28…30 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:4028-63-1
Formula:C₉H₁₁I
MDL Number:MFCD00013707