kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
6-Nitroquinoline
6-Nitroquinoline
6-Nitroquinoline
6-Nitroquinoline
Supplier:
Apollo Scientific
Certificates
Expand image
6-Nitroquinoline
Supplier:
Apollo Scientific
About this item
CAS Number:
613-50-3
:
Formula:
C₉H₆N₂O₂
MDL Number:
MFCD00006799
:
174.16 g/mol
Melting Pt:
151…153 °C (subl.)
Storage Temperature:
Ambient