Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Benzyloxycarbonyl-3-piperidinemethanol
1-Benzyloxycarbonyl-3-piperidinemethanol
1-Benzyloxycarbonyl-3-piperidinemethanol
1-Benzyloxycarbonyl-3-piperidinemethanol
Leverandør:
Apollo Scientific
Sertifikat
Expand image
1-Benzyloxycarbonyl-3-piperidinemethanol
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
39945-51-2
:
Formel:
C₁₄H₁₉NO₃
MDL-nummer:
MFCD03407298
:
249.31 g/mol
:
GHS07
Lagringstemperatur:
Ambient