
2H-Chromene-3-carboxylic acid
Catalog # APOSOR23377-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:176.17 g/mol
Melting Pt:187…189 °C
:GHS07
Storage Temperature:Ambient
CAS Number:22649-28-1
Formula:C₁₀H₈O₃
MDL Number:MFCD00202020