6-Phenoxynicotinic acid 98%
Catalog # APOSOR23316-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.2048 g/mol
Melting Pt:164…166 °C
:GHS07
Storage Temperature:Ambient
CAS Number:51362-38-0
Formula:C₁₂H₉NO₃
MDL Number:MFCD02682068