Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Furoic acid
3-Furoic acid 98%
3-Furoic acid 98%
Irritant
3-Furoic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Furoic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
488-93-7
:
Formula:
C₅H₄O₃
MDL Number:
MFCD00005350
:
112.08 g/mol
Melting Pt:
120…123 °C
EINECS:
207-689-9
:
GHS07
Storage Temperature:
Ambient