Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Diamino-6-hydroxypyrimidine
2,4-Diamino-6-hydroxypyrimidine
2,4-Diamino-6-hydroxypyrimidine
Irritant
2,4-Diamino-6-hydroxypyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Diamino-6-hydroxypyrimidine
Supplier:
Apollo Scientific
About this item
CAS Number:
56-06-4
:
MFCD08528919
Formula:
C₄H₆N₄O
MDL Number:
MFCD00006098
:
126.12 g/mol
:
GHS07
Storage Temperature:
Ambient