Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-3-nitrobenzene-1-sulphonic acid
4-Amino-3-nitrobenzene-1-sulphonic acid
4-Amino-3-nitrobenzene-1-sulphonic acid
Irritant
4-Amino-3-nitrobenzene-1-sulphonic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Amino-3-nitrobenzene-1-sulphonic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
616-84-2
:
Formula:
C₆H₆N₂O₅S
MDL Number:
MFCD00035961
:
218.19 g/mol
:
GHS07
Storage Temperature:
Ambient