Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Cinnamoylthiophene
2-Cinnamoylthiophene 98%
2-Cinnamoylthiophene 98%
2-Cinnamoylthiophene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Cinnamoylthiophene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
3988-77-0
:
Formula:
C₁₃H₁₀OS
MDL Number:
MFCD00059742
:
214.28 g/mol
:
GHS07
Storage Temperature:
Ambient