Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%
Catalog # APOSOR21663-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:222.03 g/mol
Melting Pt:227…230 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
CAS Number:1458-18-0
Formula:C₆H₅Cl₂N₃O₂
MDL Number:MFCD00010431