Order Entry
Norway
ContactUsLinkComponent
4-(tert-Butyldimethylsilyloxy)phenylboronic acid 97%
4-(tert-Butyldimethylsilyloxy)phenylboronic acid 97%
Catalog # APOSOR2113-5G
Supplier:  Apollo Scientific
CAS Number:  
4-(tert-Butyldimethylsilyloxy)phenylboronic acid 97%
Catalog # APOSOR2113-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :252.1900 g/mol
Storage Temperature:Ambient
CAS Number:159191-56-7
Formula:C₁₂H₂₁BO₃Si
MDL Number:MFCD03093888